Learn More
Med Vet International McKesson Transducer Cover, 5-1/2 X 24 Inch, 16-1401, 240/Case
Supplier:  Med Vet International 1087359CS
Encompass_Preferred
Ultrasound Transducer Cover Kit McKesson 5-1/2 X 24 Inch Polyurethane Sterile For use with Ultrasound External Probe 5½ Inches by 24 Inches Made of soft and durable polyurethane Acoustically transparent providing distortion-free imaging solutions in the ultrasound suite Designed specifically for ultrasound use to be used as a protective covering over ultrasound scanhead instruments and cord offering a sterile solution for a wide variety of transducers Slides easily over transducers and is an alternative for latex-sensitive patients and professionals Telescopically folded and straight-walled for added ease of gel insertion and cover application 24-inch length to offer additional sterile protection aiding in the prevention of cross-contamination Includes 20-mL, sterile ultrasound gel to improve conductivity during ultrasound diagnostics Includes sterile elastic band to secure transducer cover For single use only
Catalog No. 50-304-2882