Learn More
Med Vet International McKesson Ultrasound Transducer Cover Kit, 4 X 24 Inch, 16-1403, 24/Box
Catalog No. 503043660
Encompass_Preferred
Ultrasound Transducer Cover Kit McKesson 4 X 24 Inch Polyethylene Sterile For use with Ultrasound External Probe 4 x 24 in. cover Acoustically transparent, providing distortion-free imaging solutions in the ultrasound suite Telescopically folded and straight-walled for added ease of gel insertion and cover application 24-inch length to offer additional sterile protection aiding in the prevention of cross-contamination Includes 20 mL, sterile ultrasound gel to improve conductivity during ultrasound diagnostics Includes sterile elastic band to secure transducer cover Not made with natural rubber latex
Catalog No. 50-304-3660 Supplier Med Vet International Supplier No. 1087361BX
Ultrasound Transducer Cover Kit McKesson 4 X 24 Inch Polyethylene Sterile For use with Ultrasound External Probe 4 x 24 in. cover Acoustically transparent, providing distortion-free imaging solutions in the ultrasound suite Telescopically folded and straight-walled for added ease of gel insertion and cover application 24-inch length to offer additional sterile protection aiding in the prevention of cross-contamination Includes 20 mL, sterile ultrasound gel to improve conductivity during ultrasound diagnostics Includes sterile elastic band to secure transducer cover Not made with natural rubber latex