Learn More
Med Vet International McKesson Transducer Cover, 5-1/2 X 58 Inch, 16-1406, 240/Case
Catalog No. 503042953
Encompass_Preferred
Ultrasound Transducer Cover Kit McKesson 5-1/2 X 58 Inch Polyurethane Sterile For use with Ultrasound External Probe 5½ Inch by 58 Inches Made of soft and durable polyurethane Acoustically transparent providing distortion-free imaging solutions in the ultrasound suite Designed specifically for ultrasound use to be used as a protective covering over ultrasound scanhead instruments and cord offering a sterile solution for a wide variety of transducers Slides easily over transducers and is an alternative for latex-sensitive patients and professionals Telescopically folded and straight-walled for added ease of gel insertion and cover application Extra-long to offer additional sterile protection aiding in the prevention of cross-contamination Includes 20-mL, sterile ultrasound gel to improve conductivity during ultrasound diagnostics Includes colored, sterile elastic band to secure transducer cover For single use only
Catalog No. 50-304-2953 Supplier Med Vet International Supplier No. 1087363CS
Add to Cart
Add to Cart
Request a Quote
Ultrasound Transducer Cover Kit McKesson 5-1/2 X 58 Inch Polyurethane Sterile For use with Ultrasound External Probe 5½ Inch by 58 Inches Made of soft and durable polyurethane Acoustically transparent providing distortion-free imaging solutions in the ultrasound suite Designed specifically for ultrasound use to be used as a protective covering over ultrasound scanhead instruments and cord offering a sterile solution for a wide variety of transducers Slides easily over transducers and is an alternative for latex-sensitive patients and professionals Telescopically folded and straight-walled for added ease of gel insertion and cover application Extra-long to offer additional sterile protection aiding in the prevention of cross-contamination Includes 20-mL, sterile ultrasound gel to improve conductivity during ultrasound diagnostics Includes colored, sterile elastic band to secure transducer cover For single use only