Learn More
STERIS™ Coverage Plus NPD One-Step Cleaner Disinfectant
Catalog No. 17018328 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
55 gal.
Catalog No. Volume (English)
17-018-328 55 gal.
1 options
Catalog No. 17-018-328 Supplier STERIS™ Supplier No. 638701
Add to Cart
Add to Cart
Coverage Plus NPD One-Step Disinfectant is a one-step cleaner, disinfectant and deodorizer concentrate consisting of four quaternary ammonium compound chains formulated with a compatible biodegradable detergent system. pH (undiluted) - 13.0, typical
STERIS' Coverage Plus NPD One-Step Cleaner Disinfectant a phosphate-free, one-step cleaner/disinfectant formulated for use on hard non-porous surfaces. This product consists of four quaternary ammonium compounds formulated with a compatible detergent system. It is proven effective in the presence of heavy soil (5% bovine serum) and hard water. Coverage Plus NPD disinfectant contains no perfumes or dyes that could interfere with critical research studies or manufacturing processes. Use-dilution is 1:256.
HOW COVERAGE PLUS NPD ONE-STEP CLEANER DISINFECTANT WORKS
STERIS' Coverage Plus NPD One-Step Cleaner Disinfectant is a liquid concentrate specifically formulated for the disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.
WHY COVERAGE PLUS NPD ONE-STEP CLEANER DISINFECTANT?
  • Efficacy - Quaternary ammonium active ingredients provide broad spectrum antimicrobial efficacy.
  • Productivity - Formulation includes surfactants for surface cleaning while disinfecting.
  • Ease of Use - Ideal for use in a contamination control rotation.
  • Reliability - Complete lot traceability.
  • Assurance - Each lot documented by Certificate of Analysis.
Specifications
Product Type Foaming Trigger Sprayer
Color-coded No
Form Clear Liquid
Scent Unscented
Sterility Non-sterile
Size 55 gal.
Volume (English) 55 gal.