Learn More
CleanSpace™ Technology Coverall
Catalog No. 17800620 Shop All Cleanspace Technology Products
Change view
Click to view available options
Quantity:
25/Pk.
Catalog No. Quantity
17-800-620 25/Pk.
1 options
Catalog No. 17-800-620 Supplier CleanSpace™ Technology Supplier No. PAF0049
Add to Cart
Add to Cart
Disposable coverall protects CleanSpace™ respirators from dirt and splash.
  • Used with the revolutionary CleanSpace™- a light weight PAPR with no hoses/belts
  • Materials: Spun polymer fibres
  • Easy and quickly fitted and removed from the power unit
  • Compatible with all CleanSpace™ Respirators
Specifications
Disposable Reusable
Product Type Blower Cover
For Use With (Equipment) Standard and Large Case Filters
Model PAF-0049
Quantity 25/Pk.