Learn More
National Safety Apparel™ Enespro™ ArcGuard™ 12 cal Protera Coverall
SDP
Catalog No. 17001416 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
X-Large
Catalog No. Size
17-001-416 X-Large
1 options
Catalog No. 17-001-416 Supplier National Safety Apparel™ Supplier No. C88LYXL32
Coveralls with two-way covered zipper down front for easy entry
  • Constructed from Protera™
  • Comfortable, sewn-in elastic waist
  • Two-way covered zipper down front for easy entry
  • Breast pockets with flaps and hidden snap closures
  • Two large rear pockets
  • Two front pockets with pass-through slashes for access to pants pockets
  • All metal covered to ensure no contact
Specifications
Product Type Coveralls
Size X-Large
Gender Unisex
Material DuPont™ Protera™
Color Navy
Disposable Reusable
Waist Style Elastic (Stitched-in)
Ankle Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Protection Class CAT 2
Resistance Flame
Arc Flash Rating 12 cal/cm²
Certifications/Compliance ASTM F1506, NFPA 70E, CSA Z462, OSHA 1910.269
Suspender None
Style Coveralls