Learn More
Enespro™ Enespro™ ArcGuard™ 40 cal Coverall
SDP
Catalog No. 17001604 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-604 5X-Large
17-001-597 Small
17-001-598 Medium
17-001-599 Large
17-001-600 X-Large
17-001-601 2X-Large
17-001-602 3X-Large
17-001-603 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-604 Supplier Enespro™ Supplier No. C88UQUQ5X32
Protective clothing that provides the highest level of protection in work with electrical exposures and hazards
  • Recoil action allows full range of motion
  • All metal covered to ensure no skin contact
  • Chest pockets with flaps and concealed snap closures
  • Comfortable elastic sewn waist
  • Convenient side pockets for access to trouser pockets
  • FR fabric will NOT melt, drip or ignite
  • Two front pockets with step to access trouser pockets
Specifications
Product Type Coveralls
Size 5X-Large
Gender Unisex
Material Cotton, Nylon
Color Navy
Disposable Reusable
Waist Style Elastic (Stitched-in)
Ankle Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type FR Hook and Loop
Seam Type Stitched
Sterility Non-sterile
Protection Class CAT 4
Resistance Flame
Arc Flash Rating 40 cal/cm²
Certifications/Compliance NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Suspender None
Style Coveralls