Learn More
Med Vet International Non-Sterile Coverall, Extra Protection, 2X-Large, White, Disposable, 24/Case
Catalog No. 503047483
Encompass_Preferred
Back gather Offers protection from fluid strikethrough for light to moderate fluid contact 3-layer SMS fabric Non-Sterile Without Pockets Zipper Closure Elastic Wrist / Elastic Ankle Disposable
Catalog No. 50-304-7483 Supplier Med Vet International Supplier No. 165765CS
Back gather Offers protection from fluid strikethrough for light to moderate fluid contact 3-layer SMS fabric Non-Sterile Without Pockets Zipper Closure Elastic Wrist / Elastic Ankle Disposable