Learn More
Andwin Scientific COVERALL TYVEK ELAS Large, Case of 25
SDP
Catalog No. NC0621629
Encompass_Preferred
Constructed from Dupont Tyvek Isoclean material, the large Signature Coverall with elastic ankles offers a protective, durable and comfortable barrier against dry particulates, nonhazardous liquids and microorganisms.
This item is not returnable. View return policy
Catalog No. NC0621629 Supplier Andwin Scientific Supplier No. 89012826
Constructed from Dupont Tyvek Isoclean material, the large Signature Coverall with elastic ankles offers a protective, durable and comfortable barrier against dry particulates, nonhazardous liquids and microorganisms.