Learn More
Dukal™ Spunbonded Polypropylene Coveralls
Catalog No. 22415810
Change view
Click to view available options
Size:
X-Large
2X-Large
3X-Large
Color:
Blue
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
22-415-810 2X-Large White
22-415-809 X-Large White
22-415-762 X-Large Blue
22-415-761 3X-Large White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-415-810 Supplier Dukal™ Supplier No. 382XXL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Non-sterile coveralls feature a front zip closure with elastic wrists and ankle cuffs.
  • Made of durable spun bonded polypropylene.
  • Front zip closure.
  • Elastic wrist and ankle cuffs.
Specifications
Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material Spunbond Polypropylene
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance Liquid
Latex Free Yes