Learn More
Cole-Parmer Cole-Parmer Covered Container, 304 SS, 1 1/4 qt
Catalog No. NC1816675 Shop All Cole-Parmer Products
Encompass_Preferred
Highly non-corrosive 304 stainless steel (SS) can be safely heated up to 650°F (182°C). Smooth finish provides for easy cleaning.
Catalog No. NC1816675 Supplier Cole-Parmer Supplier No. 0725100
Highly non-corrosive 304 stainless steel (SS) can be safely heated up to 650°F (182°C). Smooth finish provides for easy cleaning.