Learn More
Electron Microscopy Sciences Covered Slide Warmer, 220 V
SDP
Catalog No. 501926623
Encompass_Preferred
Ideal for use in the fields of cytology, histology, pathology, and biology for parffin tissue section mounting. Thermal heater ensures even heat transfer. Anodized black surface provides contrast. LED temperature display. Size 25" x 8"
Catalog No. 50-192-6623 Supplier Electron Microscopy Sciences Supplier No. 7132120
Ideal for use in the fields of cytology, histology, pathology, and biology for parffin tissue section mounting. Thermal heater ensures even heat transfer. Anodized black surface provides contrast. LED temperature display. Size 25" x 8"