Learn More
Electron Microscopy Sciences Gold Seal 3423 Cover Glass #1.5, 24 x 60 mm, 47 pieces/oz Packaging 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 6379310
Encompass_Preferred
Made from pre-selected, pre-cleaned silicate glass. Packaged with desiccants in a lint free box.
Gold Seal: 3423
Size: 24 x 60 mm
Packaging: 47 pieces/oz
Catalog No. 50-311-32