Learn More
Electron Microscopy Sciences Gold Seal 3421 Cover Glass #1.5, 24 x 40 mm, 70 pieces/oz Packaging 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 6379101
Encompass_Preferred
Made from pre-selected, pre-cleaned silicate glass. Packaged with desiccants in a lint free box.
Gold Seal: 3421
Size: 24 x 40 mm
Packaging: 70 pieces/oz
Catalog No. 50-311-49