Learn More
Electron Microscopy Sciences Gold Seal 3419 Cover Glass #1.5, 24 x 30 mm, 94 pieces/oz Packaging 1 OZ
SDP
Catalog No. 5031151
Encompass_Preferred
Made from pre-selected, pre-cleaned silicate glass. Packaged with desiccants in a lint free box.
Gold Seal: 3419
Size: 24 x 30 mm
Packaging: 94 pieces/oz
Catalog No. 50-311-51 Supplier Electron Microscopy Sciences Supplier No. 6379001
Made from pre-selected, pre-cleaned silicate glass. Packaged with desiccants in a lint free box.
Gold Seal: 3419
Size: 24 x 30 mm
Packaging: 94 pieces/oz