Learn More
Andwin Scientific COVERGLASS #1 24X50 EA
SDP
Catalog No. 50727483
Encompass_Preferred
The Gold Seal 24X50mm with 0.13-0.17mm thickness Cover Glass is constructed from preselected and precleaned silicate glass. It comes in a lint free box that could be converted to slide storage. It is sold an ounce a piece.
This item is not returnable. View return policy
Catalog No. 50-727-483 Supplier Andwin Scientific Supplier No. 6376901
The Gold Seal 24X50mm with 0.13-0.17mm thickness Cover Glass is constructed from preselected and precleaned silicate glass. It comes in a lint free box that could be converted to slide storage. It is sold an ounce a piece.