Learn More
Andwin Scientific COVERGLASS #1.5 18X18 EA
SDP
Catalog No. 50727488
Encompass_Preferred
The Gold Seal 18 x 18mm with 0.16-0.19mm thickness Cover Glass is made from preselected and precleaned silicate glass. This product comes in a lint free box that can be converted to slide storage. This product is sold an ounce a piece.
Catalog No. 50-727-488 Supplier Andwin Scientific Supplier No. 6378701
The Gold Seal 18 x 18mm with 0.16-0.19mm thickness Cover Glass is made from preselected and precleaned silicate glass. This product comes in a lint free box that can be converted to slide storage. This product is sold an ounce a piece.