Learn More
Andwin Scientific COVERGLASS #1.5 24X50 EA
SDP
Catalog No. 50727377
Encompass_Preferred
The Gold Seal 24X50mm with 0.16-0.19mm thickness Cover Glass is constructed from preselected and precleaned silicate glass. It comes in a lint free box that could be converted to slide storage. It is sold an ounce a piece.
Catalog No. 50-727-377 Supplier Andwin Scientific Supplier No. 6379210
The Gold Seal 24X50mm with 0.16-0.19mm thickness Cover Glass is constructed from preselected and precleaned silicate glass. It comes in a lint free box that could be converted to slide storage. It is sold an ounce a piece.