Learn More
Electron Microscopy Sciences Gold Seal 3318 Cover Glass #1, 22 x 60 mm, 58 pieces/oz Packaging 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 6376510
Encompass_Preferred
Made from pre-selected, pre-cleaned silicate glass. Packaged with desiccants in a lint free box.
Gold Seal: 3318
Size: 22 x 60 mm
Packaging: 58 pieces/oz
Catalog No. 50-311-26