Learn More
Electron Microscopy Sciences Gold Seal 3321 Cover Glass #1, 24 x 40 mm, 80 pieces/oz Packaging 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 6376810
Encompass_Preferred
Made from pre-selected, pre-cleaned silicate glass. Packaged with desiccants in a lint free box.
Gold Seal: 3321
Size: 24 x 40 mm
Packaging: 80 pieces/oz
Catalog No. 50-311-20