Learn More
Electron Microscopy Sciences Gold Seal 3319 Cover Glass #1, 24 x 30 mm, 108 pieces/oz Packaging 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 6376601
Encompass_Preferred
Made from pre-selected, pre-cleaned silicate glass. Packaged with desiccants in a lint free box.
Gold Seal: 3319
Size: 24 x 30 mm
Packaging: 108 pieces/oz
Catalog No. 50-311-27