Learn More
Electron Microscopy Sciences Gold Seal 3306 Cover Glass #1, 22 x 22 mm, 156 pieces/oz Packaging 1 OZ
SDP
Catalog No. 5031133
Encompass_Preferred
Made from pre-selected, pre-cleaned silicate glass. Packaged with desiccants in a lint free box.
Gold Seal: 3306
Size: 22 x 22 mm
Packaging: 156 pieces/oz
Catalog No. 50-311-33 Supplier Electron Microscopy Sciences Supplier No. 6375701
Made from pre-selected, pre-cleaned silicate glass. Packaged with desiccants in a lint free box.
Gold Seal: 3306
Size: 22 x 22 mm
Packaging: 156 pieces/oz