Learn More
Electron Microscopy Sciences Gold Seal 3305 Cover Glass #1, 18 x 18 mm, 233 pieces/oz Packaging 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 6375610
Encompass_Preferred
Made from pre-selected, pre-cleaned silicate glass. Packaged with desiccants in a lint free box.
Gold Seal: 3305
Size: 18 x 18 mm
Packaging: 233 pieces/oz
Catalog No. 50-311-65