Learn More
Leica Biosystems COVERGLASS 24X60 540PCS
Catalog No. NC1672857
Encompass_Preferred
Premier Cover Glass has been specially designed to work with automated coverslippers. Extra processes to prevent sticking are employed during manufacturing. Premier coverglass available in multiple sizes. Thinness: 013mm - 0.17mm Sizes range from 22 X 40mm to 24 x 60mm as noted below.
Catalog No. NC1672857 Supplier Leica Biosystems Supplier No. 3800160ACS
Premier Cover Glass has been specially designed to work with automated coverslippers. Extra processes to prevent sticking are employed during manufacturing. Premier coverglass available in multiple sizes. Thinness: 013mm - 0.17mm Sizes range from 22 X 40mm to 24 x 60mm as noted below.