Learn More
Andwin Scientific COVERGLASS 24X60 EA
SDP
Catalog No. 50727480
Encompass_Preferred
The Gold Seal 24x60mm Cover Glass is constructed from preselected and precleaned silicate glass. It comes with desiccants and a lint free box that could be converted to slide storage. It is sold an ounce a piece.
Catalog No. 50-727-480 Supplier Andwin Scientific Supplier No. 6375101
The Gold Seal 24x60mm Cover Glass is constructed from preselected and precleaned silicate glass. It comes with desiccants and a lint free box that could be converted to slide storage. It is sold an ounce a piece.