Learn More
Ted Pella Inc Large Cover Glass 3 x 3-1/2" (76 x 89mm), 50/pk
SDP
Catalog No. NC0719784
Encompass
Large cover glass specially made for the large microscope slides. Made from optical grade borosilicate glass. Sizes available 3 x 3-1/4" up to 4-1/2 x 6-1/4".
Catalog No. NC0719784 Supplier Ted Pella Inc Supplier No. 260451
Large cover glass specially made for the large microscope slides. Made from optical grade borosilicate glass. Sizes available 3 x 3-1/4" up to 4-1/2 x 6-1/4".