Learn More
Electron Microscopy Sciences Gold Seal 3206 Cover Glass #0, 22 x 22 mm, 208 pieces/oz Packaging 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 6375010
Encompass_Preferred
Made from pre-selected, pre-cleaned silicate glass. Packaged with desiccants in a lint free box.
Gold Seal: 3206
Size: 22 x 22 mm
Packaging: 208 pieces/oz
Catalog No. 50-311-61