Learn More
Electron Microscopy Sciences Coverglass Staining Jar
SDP
Catalog No. 50949434
Encompass_Preferred
Coverglass staining jars for use with cover glass sizes 18 to 22 mm. Accommodates eight cover glasses back-to-back. Cap Not Included.
Catalog No. 50-949-434 Supplier Electron Microscopy Sciences Supplier No. 7224201
Coverglass staining jars for use with cover glass sizes 18 to 22 mm. Accommodates eight cover glasses back-to-back. Cap Not Included.