Learn More
Electron Microscopy Sciences Coverglass Staining Jar with Screw Cap
SDP
Supplier:  Electron Microscopy Sciences 7224221
Encompass_Preferred
For use with cover glass sizes 18 to 22 mm, accommodating up to eight back-to-back. Plastic screw caps have been tested to be resistant against most staining solutions. Comes with a plastic screw cap. Caps have been tested to be resistant against most staining solutions.
Catalog No. 50-949-372