Learn More
Electron Microscopy Sciences Coverglass Staining Jar with Glass Cap
SDP
Catalog No. 50949652
Encompass_Preferred
Coverglass staining jars for use with cover glass sizes 18 to 22 mm. Accommodates eight cover glasses back-to-back. Comes with a plastic cover. Comes with a grounded glass cover.
Catalog No. 50-949-652 Supplier Electron Microscopy Sciences Supplier No. 7224204
Coverglass staining jars for use with cover glass sizes 18 to 22 mm. Accommodates eight cover glasses back-to-back. Comes with a plastic cover. Comes with a grounded glass cover.