Learn More
Electron Microscopy Sciences Rat Tail Collagen German Glass Coverslips #1.5, 12 mm
SDP
Supplier:  Electron Microscopy Sciences 7229503
Encompass_Preferred
Coated with rat tail type I collagen on top of poly-d-lysine layer. Sterilized by radiation, ready to use. Tested for cell and neuron cultures. User manual included.
Diameter: 12 mm
Catalog No. 50-192-9592