Learn More
Electron Microscopy Sciences Fibronectin German Glass Coverslips #1, 12mm
SDP
Catalog No. 501929612
Encompass_Preferred
Coated with fibronectin on top of poly-d-lysine layer. Sterilized by radiation, ready to use. Tested for cell and neuron cultures. User manual included.
Diameter: 12mm
Catalog No. 50-192-9612 Supplier Electron Microscopy Sciences Supplier No. 7229701
Coated with fibronectin on top of poly-d-lysine layer. Sterilized by radiation, ready to use. Tested for cell and neuron cultures. User manual included.
Diameter: 12mm