Learn More
Electron Microscopy Sciences Fibronectin German Glass Coverslips #1, 20mm
SDP
Catalog No. 501929618
Encompass_Preferred
Coated with fibronectin on top of poly-d-lysine layer. Sterilized by radiation, ready to use. Tested for cell and neuron cultures. User manual included.
Diameter: 20mm
Catalog No. 50-192-9618 Supplier Electron Microscopy Sciences Supplier No. 7229707
Coated with fibronectin on top of poly-d-lysine layer. Sterilized by radiation, ready to use. Tested for cell and neuron cultures. User manual included.
Diameter: 20mm