Learn More
Electron Microscopy Sciences Gelatin German Glass Coverslips #1, 18 mm
SDP
Catalog No. 501929561
Encompass_Preferred
Sterilized by radiation, ready to use. Tested for cell cultures. User manual included.
Diameter: 18 mm
Catalog No. 50-192-9561 Supplier Electron Microscopy Sciences Supplier No. 7229305
Sterilized by radiation, ready to use. Tested for cell cultures. User manual included.
Diameter: 18 mm