Learn More
Electron Microscopy Sciences German Glass Coverslips #1, 12 mm
SDP
Catalog No. 501929516
Encompass_Preferred
CE certified crystal clear German glass coverslips for cell culture and fluorescence imaging. Tested for cell and neuron cultures. Untreated (pre-treatment required before cell culture).
Diameter: 12 mm
Catalog No. 50-192-9516 Supplier Electron Microscopy Sciences Supplier No. 7229002
CE certified crystal clear German glass coverslips for cell culture and fluorescence imaging. Tested for cell and neuron cultures. Untreated (pre-treatment required before cell culture).
Diameter: 12 mm