Learn More
Electron Microscopy Sciences German Glass Coverslips #1.5, 12 mm 0.5 OZ
SDP
Catalog No. 501929518
Encompass_Preferred
CE certified crystal clear German glass coverslips for cell culture and fluorescence imaging. Tested for cell and neuron cultures. Untreated (pre-treatment required before cell culture).
Diameter: 12 mm
Catalog No. 50-192-9518 Supplier Electron Microscopy Sciences Supplier No. 7229004
CE certified crystal clear German glass coverslips for cell culture and fluorescence imaging. Tested for cell and neuron cultures. Untreated (pre-treatment required before cell culture).
Diameter: 12 mm