Learn More
Electron Microscopy Sciences German Glass Coverslips #1.5, 18 mm 0.5 OZ
SDP
Catalog No. 501929524
Encompass_Preferred
CE certified crystal clear German glass coverslips for cell culture and fluorescence imaging. Tested for cell and neuron cultures. Untreated (pre-treatment required before cell culture).
Diameter: 18 mm
Catalog No. 50-192-9524 Supplier Electron Microscopy Sciences Supplier No. 7229010
CE certified crystal clear German glass coverslips for cell culture and fluorescence imaging. Tested for cell and neuron cultures. Untreated (pre-treatment required before cell culture).
Diameter: 18 mm