Learn More
Ibidi USA Glass Coverslips for 3 Well Chamber, removable (No. 1.5H, 15mm, Unsterile)
SDP
Catalog No. NC1181503
Encompass
Glass Coverslips for 3 Well Chamber, removable. Comes with convenient coverslip pick-up tool. No. 1.5H (170 ¿m +/- 5 ¿m) D 263 M Schott glass, ¿15 mm, unsterile, 100 pcs./box
This item is not returnable. View return policy
Catalog No. NC1181503 Supplier Ibidi USA Supplier No. 10815
Glass Coverslips for 3 Well Chamber, removable. Comes with convenient coverslip pick-up tool. No. 1.5H (170 ¿m +/- 5 ¿m) D 263 M Schott glass, ¿15 mm, unsterile, 100 pcs./box