Learn More
Electron Microscopy Sciences Coverslip Holder 1 for CPD
SDP
Catalog No. 5025139
Encompass_Preferred
Brass/nickel plated coverslip holder. Coverslips can be stacked with spacers with samples placed on top of each coverslip. Fluids freely exchange between coverslips. Both spacers and weights come with the unit. The upper part of the holder is grooved to accept tweezers. For coverslips 9 to 13mm diameter.
Catalog No. 50-251-39 Supplier Electron Microscopy Sciences Supplier No. 7019301
Brass/nickel plated coverslip holder. Coverslips can be stacked with spacers with samples placed on top of each coverslip. Fluids freely exchange between coverslips. Both spacers and weights come with the unit. The upper part of the holder is grooved to accept tweezers. For coverslips 9 to 13mm diameter.