Learn More
Electron Microscopy Sciences Coverslip Holder 2 for CPD
SDP
Catalog No. 5025138
Encompass_Preferred
Brass/nickel plated coverslip holder. Coverslips can be stacked with spacers with samples placed on top of each coverslip. Fluids freely exchange between coverslips. Both spacers and weights come with the unit. The upper part of the holder is grooved to accept tweezers. For coverslips 16 to 22mm diameter.
Catalog No. 50-251-38 Supplier Electron Microscopy Sciences Supplier No. 7019302
Brass/nickel plated coverslip holder. Coverslips can be stacked with spacers with samples placed on top of each coverslip. Fluids freely exchange between coverslips. Both spacers and weights come with the unit. The upper part of the holder is grooved to accept tweezers. For coverslips 16 to 22mm diameter.