Learn More
Electron Microscopy Sciences EMS Coverslip Holder Double
SDP
Supplier:  Electron Microscopy Sciences 72269100
Encompass_Preferred
EMS introduces an alternative to the standard aluminum slide used to visualize live microscopic organisms (both dorsal and ventral sides) using a pair of coverslips. The holders are made from refined ABS plastic and are reusable. Total slide dimensions: 3" x 1" x 0.045". Well dimensions: 25 mm x 23 mm. Inside opening dimensions: 22 mm x 16 mm. Recommended coverslip: 22 mm x 22 mm Holds coverslip sandwich in place w/out glue. Holds living material between the coverslips, generally in water.
Catalog No. 50-192-8281