Learn More
Research Products International Corp Coverslip Jar
SDP
Catalog No. 50476603
Encompass
Coverslip Jar is designed to hold up to 4 coverslips during staining applications. Ideal size jar for cleaning coverslips and polylysine/silane coating appplications. Designed to hold 17-23mm x 30mm coverslips. Longer coverslips can be accommodated if cap is removed.
Supplied with polypropylene screw cap and PTFE coated poly liner.
Catalog No. 50-476-603 Supplier Research Products International Corp Supplier No. 900180
Coverslip Jar is designed to hold up to 4 coverslips during staining applications. Ideal size jar for cleaning coverslips and polylysine/silane coating appplications. Designed to hold 17-23mm x 30mm coverslips. Longer coverslips can be accommodated if cap is removed.
Supplied with polypropylene screw cap and PTFE coated poly liner.