Learn More
Electron Microscopy Sciences Laminin German Glass Coverslips #1, 18 mm
SDP
Catalog No. 501929627
Encompass_Preferred
Coated with natural human laminin on top of poly-d-lysine layer. Sterilized by radiation, ready to use. Tested for cell and neuron cultures. User manual included.
Diameter: 18 mm
Catalog No. 50-192-9627 Supplier Electron Microscopy Sciences Supplier No. 7229805
Coated with natural human laminin on top of poly-d-lysine layer. Sterilized by radiation, ready to use. Tested for cell and neuron cultures. User manual included.
Diameter: 18 mm