Learn More
Electron Microscopy Sciences Laminin German Glass Coverslips #1, 24 mm
SDP
Supplier:  Electron Microscopy Sciences 7229811
Encompass_Preferred
Coated with natural human laminin on top of poly-d-lysine layer. Sterilized by radiation, ready to use. Tested for cell and neuron cultures. User manual included.
Diameter: 24 mm
Catalog No. 50-192-9633