Learn More
Electron Microscopy Sciences Rinzl Plastic Coverslips, 18 x 18 mm
SDP
Catalog No. 501899802
Encompass_Preferred
Plastic coverslips made from clear vinyl plastic. They are unbreakable, and will not scratch or cut fingers. Used in Biological and Medical labs, etc. Thickness of 0.01" or 0.28 mm and refractive index of glass. Economical and half the price of glass. Not autofluorescent.
Size: 18 x 18 mm
Catalog No. 50-189-9802 Supplier Electron Microscopy Sciences Supplier No. 7226118
Plastic coverslips made from clear vinyl plastic. They are unbreakable, and will not scratch or cut fingers. Used in Biological and Medical labs, etc. Thickness of 0.01" or 0.28 mm and refractive index of glass. Economical and half the price of glass. Not autofluorescent.
Size: 18 x 18 mm