Learn More
Electron Microscopy Sciences Poly-D-Lysine German Glass Coverslips #1, 14 mm
SDP
Catalog No. 501929573
Encompass_Preferred
Sterilized by radiation, ready to use. Tested for cell cultures. User manual included.
Diameter: 14 mm
Catalog No. 50-192-9573 Supplier Electron Microscopy Sciences Supplier No. 7229406
Sterilized by radiation, ready to use. Tested for cell cultures. User manual included.
Diameter: 14 mm