Learn More
Electron Microscopy Sciences Poly-D-Lysine German Glass Coverslips #1, 15 mm
SDP
Catalog No. 501929575
Encompass_Preferred
Sterilized by radiation, ready to use. Tested for cell cultures. User manual included.
Diameter: 15 mm
Catalog No. 50-192-9575 Supplier Electron Microscopy Sciences Supplier No. 7229408
Sterilized by radiation, ready to use. Tested for cell cultures. User manual included.
Diameter: 15 mm