Learn More
Electron Microscopy Sciences Poly-D-Lysine German Glass Coverslips #1.5, 22 mm
SDP
Catalog No. 501929585
Encompass_Preferred
Sterilized by radiation, ready to use. Tested for cell cultures. User manual included.
Diameter: 22 mm
Catalog No. 50-192-9585 Supplier Electron Microscopy Sciences Supplier No. 7229418
Sterilized by radiation, ready to use. Tested for cell cultures. User manual included.
Diameter: 22 mm