Learn More
Electron Microscopy Sciences Poly-L-Lysine German Glass Coverslips #1.5, 18 mm
SDP
Supplier:  Electron Microscopy Sciences 7229209
Encompass_Preferred
CE certified crystal clear German glass coverslips sterilized by radiation and ready to use. Tested for cell and neuron cultures. User manual included.
Diameter: 18 mm
Catalog No. 50-192-9546