Learn More
Chemglass Life Sciences Cover Slip, Rectangular, #1.5 Corning 0211 Glass, 24 x 40mm, 1oz/Pkg, Approx 71/Oz
SDP
Catalog No. 501532238
Encompass_Preferred
Cover Slip, Rectangular, #1.5 Corning 0211 Glass, 24 x 40mm, 1oz/Pkg, Approx 71/Oz. Cover slips are manufactured from Corning 0211 Float Glass, #1.5 thickness (0.16 to 0.19mm). Sold in packages of 1oz.
Catalog No. 50-153-2238 Supplier Chemglass Life Sciences Supplier No. CLS17642440
Cover Slip, Rectangular, #1.5 Corning 0211 Glass, 24 x 40mm, 1oz/Pkg, Approx 71/Oz. Cover slips are manufactured from Corning 0211 Float Glass, #1.5 thickness (0.16 to 0.19mm). Sold in packages of 1oz.